Methyl 3-chloro-4-methylthiophene-2-carboxylate

Lieferant: Apollo Scientific

OR24319-5G
APOSOR24319-5GEA 58 EUR
APOSOR24319-5G
Methyl 3-chloro-4-methylthiophene-2-carboxylate
Methyl 3-chloro-4-methylthiophene-2-carboxylate
Formel: C₇H₇ClO₂S
Molecular Weight: 190.647 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052072
CAS-Nummer: 175137-11-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR