3-Chloro-4-methylthiophene-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR24320-100MG
APOSOR24320-100MGEA 46.4 EUR
APOSOR24320-100MG
3-Chloro-4-methylthiophene-2-carbohydrazide
3-Chloro-4-methylthiophene-2-carbohydrazide
Formel: C₆H₇ClN₂OS
Molecular Weight: 190,651 g/mol
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068136
CAS-Nummer: 175137-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR