1-Phenyl-5-propyl-1H-pyrazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR24326-1G
APOSOR24326-1GEA 125 EUR
APOSOR24326-1G
1-Phenyl-5-propyl-1H-pyrazole-4-carboxylic acid
1-Phenyl-5-propyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₃H₁₄N₂O₂
Molecular Weight: 230,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00068140
CAS-Nummer: 116344-17-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR