4-Chloro-7-methylthieno[3,2-d]pyrimidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR24331-1G
APOSOR24331-1GEA
36.4
EUR
APOSOR24331-1G
4-Chloro-7-methylthieno[3,2-d]pyrimidine 97%
4-Chloro-7-methylthieno[3,2-d]pyrimidine
Formel:
C₇H₅ClN₂S Schmelzpunkt: 122…125 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00205202 CAS-Nummer: 175137-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...