4-Hydrazino-7-methylthieno[3,2-d]pyrimidine

Lieferant: Apollo Scientific

OR24332-1G
APOSOR24332-1GEA 393 EUR
APOSOR24332-1G
4-Hydrazino-7-methylthieno[3,2-d]pyrimidine
4-Hydrazino-7-methylthieno[3,2-d]pyrimidine
Formel: C₇H₈N₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00205196
CAS-Nummer: 175137-22-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR