Methyl-4-aminothiophene-3-carboxylate hydrochloride 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-aminothiophene-3-carboxylate hydrochloride

OR24349-1G
APOSOR24349-1GEA 21.9 EUR
APOSOR24349-1G
Methyl-4-aminothiophene-3-carboxylate hydrochloride 96%
Methyl-4-aminothiophene-3-carboxylate hydrochloride
Formel: C₆H₈ClNO₂S
Molecular Weight: 193.65 g/mol
Schmelzpunkt: 183…185 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00068149
CAS-Nummer: 39978-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR