3-Aminothieno[3,2-d]pyrimidin-4(3H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-3,4-dihydro-4-oxothieno[3,2-d]pyrimidine

OR24373-250MG
APOSOR24373-250MGEA 174 EUR
APOSOR24373-250MG
3-Aminothieno[3,2-d]pyrimidin-4(3H)-one
3-Aminothieno[3,2-d]pyrimidin-4(3H)-one
Formel: C₆H₅N₃OS
Molecular Weight: 167,188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00816913
CAS-Nummer: 648859-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR