(2,4-Dichlorophenoxy)acetyl chloride

Lieferant: Apollo Scientific
Gefahr

OR2440-100MG
APOSOR2440-100MGEA 23.2 EUR
APOSOR2440-100MG
(2,4-Dichlorophenoxy)acetyl chloride
(2,4-Dichlorophenoxy)acetyl chloride
Formel: C₈H₅Cl₃O₂
Molecular Weight: 239,483 g/mol
Siedepunkt: 182 °C (15 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00045225
CAS-Nummer: 774-74-3
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR