Morpholine-4-carbohydrazide

Lieferant: Apollo Scientific

OR24403-1G
APOSOR24403-1GEA 277 EUR
APOSOR24403-1G
Morpholine-4-carbohydrazide
Morpholine-4-carbohydrazide
Formel: C₅H₁₁N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01570142
CAS-Nummer: 29053-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR