1-(3-Aminoprop-1-yl)-5-(tert-butyl)-2-methyl-1H-pyrrole-3-carboxylic acid, 1-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-{3-[(tert-Butoxycarbonyl)amino]prop-1-yl}-5-(tert-butyl)-2-methyl-1H-pyrrole-3-carboxylic acid, 3-[5-(tert-Butyl)-3-carboxy-2-methyl-1H-pyrrol-1-yl]propylamine, N-BOC protected

OR2441-250MG
APOSOR2441-250MGEA 198 EUR
APOSOR2441-250MG
1-(3-Aminoprop-1-yl)-5-(tert-butyl)-2-methyl-1H-pyrrole-3-carboxylic acid, 1-BOC protected
1-(3-Aminoprop-1-yl)-5-(tert-butyl)-2-methyl-1H-pyrrole-3-carboxylic acid, 1-BOC protected
Formel: C₁₈H₃₀N₂O₄
Molecular Weight: 338,442 g/mol
Schmelzpunkt: 154…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01567234
CAS-Nummer: 306936-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR