Bicyclo[3.3.1]nonane-2,6-dione 97%

Lieferant: Apollo Scientific

OR2442-1G
APOSOR2442-1GEA 198 EUR
APOSOR2442-1G
Bicyclo[3.3.1]nonane-2,6-dione 97%
Bicyclo[3.3.1]nonane-2,6-dione
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Siedepunkt: 180 °C/20mmHg
Schmelzpunkt: 151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00153926
CAS-Nummer: 16473-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR