1-(3-Phenyl-[1,2,4]-thiadiazol-5-yl)piperazine
Lieferant: Apollo Scientific
|
Gefahr
|
OR24470-1G
APOSOR24470-1GEA
184
EUR
APOSOR24470-1G
1-(3-Phenyl-[1,2,4]-thiadiazol-5-yl)piperazine
1-(3-Phenyl-[1,2,4]-thiadiazol-5-yl)piperazine
Formel:
C₁₂H₁₄N₄S Molecular Weight: 246,34 g/mol Schmelzpunkt: 93…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00115115 CAS-Nummer: 306935-14-8 UN: 3259 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...