1,4-Diiod-2,5-dimethoxybenzol
Lieferant: Apollo Scientific
|
Achtung
|
OR2448-1G
APOSOR2448-1GEA
21.9
EUR
APOSOR2448-1G
1,4-Diiod-2,5-dimethoxybenzol
1,4-Diiodo-2,5-dimethoxybenzene
Formel:
C₈H₈I₂O₂ Molecular Weight: 389,9569 g/mol Schmelzpunkt: 171…173 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00461369 CAS-Nummer: 51560-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...