Ethyl-3-(4-chlorophenyl)-2-cyanoacrylate
Lieferant: Apollo Scientific
|
Achtung
|
OR24494-1G
APOSOR24494-1GEA
0
EUR
APOSOR24494-1G
Ethyl-3-(4-chlorophenyl)-2-cyanoacrylate
Ethyl-3-(4-chlorophenyl)-2-cyanoacrylate
Formel:
C₁₂H₁₀ClNO₂ Schmelzpunkt: 137…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00454124 CAS-Nummer: 2286-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...