Chloromethyl-4-nitrophenyl carbonate

Lieferant: Apollo Scientific

OR24520-1G
APOSOR24520-1GEA 393 EUR
APOSOR24520-1G
Chloromethyl-4-nitrophenyl carbonate
Chloromethyl-4-nitrophenyl carbonate
Formel: C₈H₆ClNO₅
Molecular Weight: 231,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00208111
CAS-Nummer: 50780-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR