7-methylchroman-4-one 95+%

Lieferant: Apollo Scientific

Synonyme: 7-Methylchroman-4-one

OR24523-250MG
APOSOR24523-250MGEA 85.7 EUR
APOSOR24523-250MG
7-methylchroman-4-one 95+%
7-methylchroman-4-one
Formel: C₁₀H₁₀O₂
Molecular Weight: 162.185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00208231
CAS-Nummer: 18385-69-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR