N-Benzyl-2-chloro-N-(2-chloroethyl)ethanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzyl[bis(2-chloroethyl)]amine hydrochloride

OR24559-250MG
APOSOR24559-250MGEA 37.7 EUR
APOSOR24559-250MG
N-Benzyl-2-chloro-N-(2-chloroethyl)ethanamine hydrochloride
N-Benzyl-2-chloro-N-(2-chloroethyl)ethanamine hydrochloride
Formel: C₁₁H₁₆Cl₃N
Molecular Weight: 268,62 g/mol
Schmelzpunkt: 134…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00185654
CAS-Nummer: 10429-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR