2-[(Phenylsulphonyl)methyl]benzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(Bromomethyl)-2-[(phenylsulphonyl)methyl]benzene

OR24578-250MG
APOSOR24578-250MGEA 21.9 EUR
APOSOR24578-250MG
2-[(Phenylsulphonyl)methyl]benzyl bromide 97%
2-[(Phenylsulphonyl)methyl]benzyl bromide
Formel: C₁₄H₁₃BrO₂S
Molecular Weight: 325,221 g/mol
Schmelzpunkt: 93…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007552
CAS-Nummer: 88116-02-3
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR