O-tert-Butylhydroxylamin Hydrochlorid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: O-(tert-Butyl)hydroxylamine hydrochloride

OR24583-25G
APOSOR24583-25GEA 121 EUR
APOSOR24583-25G
O-tert-Butylhydroxylamin Hydrochlorid 98%
O-tert-Butylhydroxylamin Hydrochlorid
Formel: C₄H₁₂ClNO
Molecular Weight: 125.5972 g/mol
Schmelzpunkt: 155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043272
CAS-Nummer: 39684-28-1
EINECS: 254-590-1
UN: 1325
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR