5-[(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-2-furoic acid

Lieferant: Apollo Scientific
Gefahr

OR2459-1G
APOSOR2459-1GEA 68.3 EUR
APOSOR2459-1G
5-[(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-2-furoic acid
5-[(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-2-furoic acid
Formel: C₁₁H₁₁BrN₂O₃
Schmelzpunkt: 207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00209309
CAS-Nummer: 306935-28-4
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR