3-Formyl-2,2,5,5-tetramethyl-1,3-thiazolidine-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Carboxy-3-formyl-2,2,5,5-tetramethyl-1,3-thiazolidine, 3-Formyl-2,2,5,5-tetramethyl-1,3-thiazolane-4-carboxylic acid

OR24610-250MG
APOSOR24610-250MGEA 56.7 EUR
APOSOR24610-250MG
3-Formyl-2,2,5,5-tetramethyl-1,3-thiazolidine-4-carboxylic acid
3-Formyl-2,2,5,5-tetramethyl-1,3-thiazolidine-4-carboxylic acid

Chiral

Formel: C₉H₁₅NO₃S
Molecular Weight: 217,285 g/mol
Schmelzpunkt: 179…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03939696
CAS-Nummer: 55234-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR