3-Bromcarbazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-9H-carbazole

OR24631-25G
APOSOR24631-25GEA 21.9 EUR
APOSOR24631-25G
3-Bromcarbazol
3-Bromocarbazole
Formel: C₁₂H₈BrN
Molecular Weight: 246,11 g/mol
Schmelzpunkt: 196…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00222621
CAS-Nummer: 1592-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR