4-Cyano-3-(4-methoxyphenyl)-5-(methylthio)thiophene-2-carboxylic acid

Lieferant: Apollo Scientific

OR2466-1G
APOSOR2466-1GEA 0 EUR
APOSOR2466-1G
4-Cyano-3-(4-methoxyphenyl)-5-(methylthio)thiophene-2-carboxylic acid
4-Cyano-3-(4-methoxyphenyl)-5-(methylthio)thiophene-2-carboxylic acid
Formel: C₁₄H₁₁NO₃S₂
Schmelzpunkt: 209…213 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00277044
CAS-Nummer: 116493-07-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR