Methyl 4-(1-hydroxyethyl)benzoate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[4-(Methoxycarbonyl)phenyl]ethan-1-ol, 4-(Methoxycarbonyl)-α-methylbenzyl alcohol

OR24665-1G
APOSOR24665-1GEA 230 EUR
APOSOR24665-1G
Methyl 4-(1-hydroxyethyl)benzoate 95%
Methyl 4-(1-hydroxyethyl)benzoate
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.203 g/mol
Siedepunkt: 148…152 °C (1 mm Hg)
Dichte: 1.44 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00216476
CAS-Nummer: 84851-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR