1-methylhexahydropyrimidine-2,4-dione
Lieferant: Apollo Scientific
OR24703-1G
APOSOR24703-1GEA
404
EUR
APOSOR24703-1G
1-methylhexahydropyrimidine-2,4-dione
1-methylhexahydropyrimidine-2,4-dione
Formel:
C₅H₈N₂O₂ Molecular Weight: 128,129 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00127872 CAS-Nummer: 696-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...