3-Benzothiazol-2-ylpropionic acid

Lieferant: Apollo Scientific
Achtung

OR2471-1G
APOSOR2471-1GEA 145 EUR
APOSOR2471-1G
3-Benzothiazol-2-ylpropionic acid
3-Benzothiazol-2-ylpropionic acid
Formel: C₁₀H₉NO₂S
Molecular Weight: 207,25 g/mol
Schmelzpunkt: 108…110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01764690
CAS-Nummer: 29198-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR