N-(Fur-2-ylmethyl)-4-methylaniline
Lieferant: Apollo Scientific
|
Achtung
|
OR24759-100MG
APOSOR24759-100MGEA
75.4
EUR
APOSOR24759-100MG
N-(Fur-2-ylmethyl)-4-methylaniline
N-(Fur-2-ylmethyl)-4-methylaniline
Formel:
C₁₂H₁₃NO Molecular Weight: 187,238 g/mol Siedepunkt: 155…157 °C (10 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00830575 CAS-Nummer: 3139-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...