4-(4-Benzylpiperazino)benzenecarbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR2478-250MG
APOSOR2478-250MGEA
0
EUR
APOSOR2478-250MG
4-(4-Benzylpiperazino)benzenecarbaldehyde
4-(4-Benzylpiperazino)benzenecarbaldehyde
Formel:
C₁₈H₂₀N₂O Schmelzpunkt: 73…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01765492 CAS-Nummer: 166438-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...