Ethyl-4-hydroxy-3-methoxycinnamat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydroxy-3-methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-hydroxy-3-methoxyphenyl)acrylate, Ethyl Ferulate, Ferulic Acid Ethyl Ester

OR24814-25G
APOSOR24814-25GEA 26.1 EUR
APOSOR24814-25G
Ethyl-4-hydroxy-3-methoxycinnamat 98%
Ethyl-4-hydroxy-3-methoxycinnamate
Formel: C₁₂H₁₄O₄
Molecular Weight: 222,24 g/mol
Siedepunkt: 164…166 °C/0,5mmHg
Schmelzpunkt: 63…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009190
CAS-Nummer: 4046-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR