1-Methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one
Lieferant: Apollo Scientific
|
Achtung
|
OR2482-100MG
APOSOR2482-100MGEA
21.9
EUR
APOSOR2482-100MG
1-Methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one
1-Methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one
Formel:
C₆H₆N₄O Schmelzpunkt: 290…292 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01312343 CAS-Nummer: 5334-56-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...