3-[4-(Benzyloxy)phenyl]acrylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 4-(Benzyloxy)cinnamic acid

OR24876-1G
APOSOR24876-1GEA 60.9 EUR
APOSOR24876-1G
3-[4-(Benzyloxy)phenyl]acrylic acid 98%
3-[4-(Benzyloxy)phenyl]acrylic acid
Formel: C₁₆H₁₄O₃
Molecular Weight: 254.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00021778
CAS-Nummer: 6272-45-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR