2-Brom-9-fluorenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-9H-fluoren-9-one

OR2491-1G OR2491-100G OR2491-500G
APOSOR2491-1GEA 21.9 EUR
APOSOR2491-1G APOSOR2491-100G APOSOR2491-500G
2-Brom-9-fluorenon
2-Brom-9-fluorenon
Formel: C₁₃H₇BrO
Molecular Weight: 259.1 g/mol
Schmelzpunkt: 146…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010789
CAS-Nummer: 3096-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR