2-Brom-9-fluorenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-9H-fluoren-9-one
OR2491-1G
OR2491-100G
OR2491-500G
APOSOR2491-1GEA
21.9
EUR
APOSOR2491-1G
APOSOR2491-100G
APOSOR2491-500G
2-Brom-9-fluorenon
2-Brom-9-fluorenon
Formel:
C₁₃H₇BrO Molecular Weight: 259.1 g/mol Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010789 CAS-Nummer: 3096-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...