2,3-Dimethylbenzyl alcohol 95+%
Lieferant: Apollo Scientific
Synonyme:
(2,3-Dimethylphenyl)methanol
OR24912-5G
APOSOR24912-5GEA
63.8
EUR
APOSOR24912-5G
2,3-Dimethylbenzyl alcohol 95+%
2,3-Dimethylbenzyl alcohol
Formel:
C₉H₁₂O Molecular Weight: 136,19 g/mol Siedepunkt: 128…131 °C/13mmHg Schmelzpunkt: 63…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046742 CAS-Nummer: 13651-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...