4-(4-Methanesulphonylpiperazin-1-yl)phenylamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(4-(Methylsulphonyl)piperazin-1-yl)aniline, 4-[4-(Methylsulphonyl)piperazin-1-yl]aniline
OR2492-1G
APOSOR2492-1GEA
126
EUR
APOSOR2492-1G
4-(4-Methanesulphonylpiperazin-1-yl)phenylamine
4-(4-Methanesulphonylpiperazin-1-yl)phenylamine
Formel:
C₁₁H₁₇N₃O₂S Molecular Weight: 255,34 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03848236 CAS-Nummer: 442549-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...