5-Amino-2-methylbenzoesäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-methylbenzoic Acid, 2-Methyl-5-aminobenzoic acid, 5-Amino-o-toluic acid

OR2497-25G
APOSOR2497-25GEA 52.2 EUR
APOSOR2497-25G
5-Amino-2-methylbenzoesäure
5-Amino-2-methylbenzoic acid
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
Schmelzpunkt: 194…199 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06208351
CAS-Nummer: 2840-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR