4-(4-Acetylpiperazin-1-yl)benzoic acid

Lieferant: Apollo Scientific

OR2498-1G OR2498-250MG OR2498-500MG OR2498-2.5G OR2498-5G
APOSOR2498-1GEA 0 EUR
APOSOR2498-1G APOSOR2498-250MG APOSOR2498-500MG APOSOR2498-2.5G APOSOR2498-5G
4-(4-Acetylpiperazin-1-yl)benzoic acid
4-(4-Acetylpiperazin-1-yl)benzoic acid
Formel: C₁₃H₁₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05862631
CAS-Nummer: 104080-55-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR