2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one

Lieferant: Apollo Scientific
Achtung

OR25017-1G
APOSOR25017-1GEA 72.5 EUR
APOSOR25017-1G
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one
Formel: C₈H₉N₃O
Molecular Weight: 163,18 g/mol
Schmelzpunkt: 252…254 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00210153
CAS-Nummer: 98488-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR