(4-Chlorphenylthio)acetonitril
Lieferant: Apollo Scientific
|
Achtung
|
OR25038-1G
OR25038-5G
OR25038-25G
APOSOR25038-1GEA
21.9
EUR
APOSOR25038-1G
APOSOR25038-5G
APOSOR25038-25G
(4-Chlorphenylthio)acetonitril
(4-Chlorphenylthio)acetonitril
Formel:
C₈H₆ClNS Schmelzpunkt: 84…90 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00019832 CAS-Nummer: 18527-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...