5-Bromo-4-methoxythiophene-3-carboxylic acid

Lieferant: Apollo Scientific

OR25051-1G
APOSOR25051-1GEA 0 EUR
APOSOR25051-1G
5-Bromo-4-methoxythiophene-3-carboxylic acid
5-Bromo-4-methoxythiophene-3-carboxylic acid
Formel: C₆H₅BrO₃S
Lagertemperatur: Raumtemperatur
CAS-Nummer: 162848-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR