3-Ethoxy-2-[(naphth-2-yl)sulphonyl]acrylonitrile

Lieferant: Apollo Scientific

OR25090-1G OR25090-5G
APOSOR25090-1GEA 87 EUR
APOSOR25090-1G APOSOR25090-5G
3-Ethoxy-2-[(naphth-2-yl)sulphonyl]acrylonitrile
3-Ethoxy-2-[(naphth-2-yl)sulphonyl]acrylonitrile
Formel: C₁₅H₁₃NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00067880
CAS-Nummer: 32083-61-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR