2-Propanesulphonylacetonitrile

Lieferant: Apollo Scientific
Achtung

OR25142-1G OR25142-250MG
APOSOR25142-1GEA 134 EUR
APOSOR25142-1G APOSOR25142-250MG
2-Propanesulphonylacetonitrile
2-Propanesulphonylacetonitrile
Formel: C₅H₉NO₂S
Molecular Weight: 147.2 g/mol
Schmelzpunkt: 44…46 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 120069-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR