Methyl 3-amino-4-(methylsulphonyl)thiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR25165-1G
APOSOR25165-1GEA 102 EUR
APOSOR25165-1G
Methyl 3-amino-4-(methylsulphonyl)thiophene-2-carboxylate
Methyl 3-amino-4-(methylsulphonyl)thiophene-2-carboxylate
Formel: C₇H₉NO₄S₂
Molecular Weight: 235,281 g/mol
Schmelzpunkt: 145…147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067932
CAS-Nummer: 175201-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR