5-(tert-butylsulphonyl)-2-methylpyrimidin-4-amine

Lieferant: Apollo Scientific

OR25182-1G
APOSOR25182-1GEA 143 EUR
APOSOR25182-1G
5-(tert-butylsulphonyl)-2-methylpyrimidin-4-amine
5-(tert-butylsulphonyl)-2-methylpyrimidin-4-amine
Formel: C₉H₁₅N₃O₂S
Molecular Weight: 229,299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067946
CAS-Nummer: 175201-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR