Ethyl 2-methyl-7-oxo-4,7-dihydropyrazolo[1,5-a]pyrimidine-6-carboxylate

Lieferant: Apollo Scientific
Achtung

OR25189-50MG
APOSOR25189-50MGEA 102 EUR
APOSOR25189-50MG
Ethyl 2-methyl-7-oxo-4,7-dihydropyrazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl 2-methyl-7-oxo-4,7-dihydropyrazolo[1,5-a]pyrimidine-6-carboxylate
Formel: C₁₀H₁₁N₃O₃
Molecular Weight: 221,213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067951
CAS-Nummer: 99056-35-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR