(S)-Cotinine N-Oxide

Lieferant: Apollo Scientific

Synonyme: (5S)-1-Methyl-5-(1-oxidopyridin-3-yl)pyrrolidin-2-one

OR2520-25MG
APOSOR2520-25MGEA 480 EUR
APOSOR2520-25MG
(S)-Cotinine N-Oxide
(S)-Cotinine N-Oxide

A metabolite of nicotine in humans.

Formel: C₁₀H₁₂N₂O₂
Molecular Weight: 192,214 g/mol
Schmelzpunkt: 116…117 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD13176618
CAS-Nummer: 36508-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR