Ethyl 1,3-dimethyl-4-(4-methoxyphenoxy)-1H-pyrazolo[3,4-b]pyridine-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-Dimethyl-5-(ethoxycarbonyl)-4-(4-methoxyphenoxy)-1H-pyrazolo[3,4-b]pyridine, 4-{[1,3-Dimethyl-5-(ethoxycarbonyl)-1H-pyrazolo[3,4-b]pyridin-4-yl]oxy}anisole
OR25206-250MG
APOSOR25206-250MGEA
69.6
EUR
APOSOR25206-250MG
Ethyl 1,3-dimethyl-4-(4-methoxyphenoxy)-1H-pyrazolo[3,4-b]pyridine-5-carboxylate
Ethyl 1,3-dimethyl-4-(4-methoxyphenoxy)-1H-pyrazolo[3,4-b]pyridine-5-carboxylate
Formel:
C₁₈H₁₉N₃O₄ Molecular Weight: 341,361 g/mol Schmelzpunkt: 92…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067964 CAS-Nummer: 174842-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...