Ethyl 3-(isopropylthio)-6,6-dimethyl-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate
Lieferant: Apollo Scientific
OR25272-1G
OR25272-5G
APOSOR25272-1GEA
72.5
EUR
APOSOR25272-1G
APOSOR25272-5G
Ethyl 3-(isopropylthio)-6,6-dimethyl-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate
Ethyl 3-(isopropylthio)-6,6-dimethyl-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate
Formel:
C₁₆H₂₂O₃S₂ Molecular Weight: 326.474 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085069 CAS-Nummer: 175202-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...