6,6-Dimethyl-3-(methylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR25283-1G
APOSOR25283-1GEA 157 EUR
APOSOR25283-1G
6,6-Dimethyl-3-(methylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carbonitrile
6,6-Dimethyl-3-(methylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carbonitrile
Formel: C₁₂H₁₃NOS₂
Molecular Weight: 251,368 g/mol
Schmelzpunkt: 169…171 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00084980
CAS-Nummer: 175202-50-3
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR