3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid

OR25290-250MG
APOSOR25290-250MGEA 87 EUR
APOSOR25290-250MG
3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid 95+%
3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid
Formel: C₁₀H₈O₄S₂
Molecular Weight: 256,298 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00067989
CAS-Nummer: 175202-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR